Acid surrogate mix (4/89 SOW)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
33029 31025.15 31025.25 31063.15
REST33029EA 310.5 EUR InStock
REST33029 REST31025.15 REST31025.25 REST31063.15
Acid surrogate mix (4/89 SOW)
Standards Umweltanalyse-Standards
Standards are packaged in methanol in an ampoule.
  • Convenient 1 ml, 5 ml and 10 ml package sizes available
  • Highest concentrations commercially available
  • Reduces laboratory cost per sample extract
  • Mixture contains 2-Fluorophenol, phenol-d6, and 2,4,6-tribromophenol
Lieferumfang: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge