GC column connectors, Defined PressFit
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
22161 22164
REST22161EA 97 EUR InStock
REST22161 REST22164
GC column connectors, Defined PressFit
Fittings und Verbinder
These PressFit connectors feature ID-specific ends for better sealability and a defined fit.
  • Deactivated borosilicate glass
  • Connect a guard column to an analytical column
  • Repair a broken column
  • Connect a column outlet to a transfer line
These connectors are colour coded for easy identification.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge