Antibody diluent clear
Lieferant: IMMUNOLOGIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
BD09-125 BD09-500 BD09-999 S/BD09-20
BD09-125EA 57.8 EUR InStock
BD09-125 BD09-500 BD09-999 IMMLS/BD09-20
Antibody diluent clear
Elektrophorese-Reagenzien Western Blotting Reagents
Universal IHC Blocking/Diluent is specially formulated to stabilise antibodies and eliminate backgrounds encountered in immunohistochemistry staining procedures. The diluent contains complex proteins and non-protein mixtures which eliminate histostaining background by blocking Fc receptors and prevents non-specific protein-protein interactions.
  • High-quality diluent solution for IHC apllications
Universal IHC Blocking/Diluent can also be used as blocking applied before primary antibodies. If primary antibodies are diluted in this diluent, a blocking step before primary antibodies can generally be ommited. The blocking/diluent solution is supplied in ready-to-use format. It should not be diluted to be effective. The diluent contains no azide, therefore, it can be used to dilute and stabilise HRP-conjugates as well as any other solution that need and stabilising carrier protein, such as post-antibody blocking and poly-AP conjugates.
Zertifizierungen: Meets CE/IVD standards
Bestellinformation: Colour version also available (see IMMLS/UD09-20 and similar products)
Lieferumfang: Shipped with blue ice. Store the products between 2 to 8°C.
Achtung! Bitte beachten: Irritating to eyes, respiratory systems and skin
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..