Bovine gamma-globulin (from Blood)
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
G5009-100G G5009-10G G5009-1G G5009-25G G5009-5G
SIALG5009-100GEA 3640 EUR InStock
SIALG5009-100G SIALG5009-10G SIALG5009-1G SIALG5009-25G SIALG5009-5G
Bovine gamma-globulin (from Blood)
Proteine und Peptide
γ-Globulin from bovine blood are categorised by anion exchange chromatography, immunoelectrophoresis, zone electrophoresis, ultracentrifugation, and analysis of the products of papain digestion. It is used as an immunological challenge in studies of mechanisms of immunotolerance.
Bovine γ-globulins are used to study the constituents and pH changes in protein rich hyaluronan solution affecting the bio-tribological properties of artificial articular joints. It is also used to determine immunoglobulin G in bovine colostrum and milk by direct biosensor SPR-immunoassay and in rapid screening for monensin residues in poultry plasma by a dry reagent dissociation enhanced lanthanide fluoroimmunoassay.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge