Rapid Gold BCA protein assay kit, Pierce™
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
A53225 A53226
A53225EA 261 EUR InStock
A53225 A53226
Rapid Gold BCA protein assay kit, Pierce™
Tests Proteintests
Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit is a two-component, high precision, detergent-compatible assay optimised to measure (at 480 nm) total protein concentration compared to a protein standard curve of known concentrations. The Pierce™ Rapid Gold BCA Protein Assay uses the same copper-chelating technology as the well-known traditional Pierce™ BCA Protein Assay and provides comparable accuracy but with a 5 minute, room temperature incubation and measured at 480 nm. This improvement eliminates the need to wait or expose the samples to elevated temperatures for a fast time to results.
  • Colorimetric – estimate visually or measure with a standard spectrophotometer or plate reader (480 nm)
  • Excellent uniformity – exhibits similar protein-to-protein variation as traditional BCA Protein Assay, and less protein-to-protein variation than dye-binding protein assay methods
  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents
  • Fast time to results – significant improvement over traditional BCA Assay with a 5 minute, room temperature reaction
  • High linearity – linear working range for BSA equals 20 to 2000 µg/mL
  • Optimal absorbance on standard spectrophotometers – reaction produces a colour change to an intense orange-gold that results in a strong linear response at 480 nm
Applications:
Studying protein:protein interactions
Measuring column fractions after affinity chromatography
Estimating percent recovery of membrane proteins from cell extracts
High-throughput screening of fusion proteins
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge