LC columns, AdvanceBio RP-mAb, Diphenyl
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
793775-944 793975-944 795775-944 795975-944 797775-944 799775-944 799975-944
HEWL793775-944EA 1166 EUR InStock
HEWL793775-944 HEWL793975-944 HEWL795775-944 HEWL795975-944 HEWL797775-944 HEWL799775-944 HEWL799975-944
LC columns, AdvanceBio RP-mAb, Diphenyl
Chromatographiesäulen
AdvanceBio RP-mAb columns are designed to optimise the performance of intact and reduced mAb analysis when analysing monoclonal antibodies for biopharma discovery, development, and QA/QC applications.
  • Superficially porous particles (3,5 µm) with wide pores (450 Å) increase mAb resolution while maintaining compatibility with all LC instruments
  • Shorter analysis times compared to columns packed with fully porous particles of the same size
  • Robust poroshell packed bed and 2 µm inlet frit extend column lifetime by helping prevent inlet blockage
The wide-pore poroshell technology of the column delivers high efficiency, a short analysis time, and low pressure, at temperatures below 80 °C.
Bestellinformation: Available in a range of chemistries, SB-C8, C4, and diphenyl.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge