Gel filtration chromatography columns, HiScale™ Superdex™ Increase and Superose™ Increase
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
29-3219-03 29-3219-04 29-3219-05 29-3219-06 29-3219-07 29-3219-08
29-3219-03EA 6877 EUR InStock
29-3219-03 29-3219-04 29-3219-05 29-3219-06 29-3219-07 29-3219-08
Gel filtration chromatography columns, HiScale™ Superdex™ Increase and Superose™ Increase
Chromatographiesäulen
These HiScale columns packed with Superdex Increase or Superose Increase resins are used to purify larger volumes of up to 8.5 ml.
  • High-resolution SEC columns: Small particle size and narrow particle size distribution of the packed resin ensures highly pure protein
  • HiScale columns packed with SEC resins on demand, please request a quote
HiScale columns are packed on demand for you with Superdex 200 Increase, Superdex 75 Increase, and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes: 16 × 400 mm and 26 × 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 ml, respectively.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge