MS spent media analysis columns, AdvanceBio
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
673775-901 675775-901 679775-901
HEWL673775-901EA 1612 EUR InStock
HEWL673775-901 HEWL675775-901 HEWL679775-901
MS spent media analysis columns, AdvanceBio
Chromatographiesäulen
These columns are suitable for fast, reproducible analysis of bioreactor cell culture media.
  • Performance: PEEK-lined stainless steel columns deliver excellent peak shape and recovery of challenging ionic metabolites with pressure tolerance up to 600 bar
  • Sensitivity: MS-friendly mobile phases provide excellent analytical sensitivity
  • Flexibility: Compatible with both HPLC and UHPLC systems via 2,7 µm Poroshell particles
AdvanceBio columns make it easier for your biotech lab to analyse amino acids and other small metabolites in spent cell culture media - with or without sample derivatisation.
Columns for both solutions are tested with amino acids to ensure quality and performance.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge