Safety spectacles, Marcus Grönholm™
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MMMAMGRO11BREA 24.7 EUR InStock
MMMAMGRO11BR MMMAMGRO3BR
Safety spectacles, Marcus Grönholm™
These safety glasses are designed around the principals of protection, style and comfort. Adjustable nose pads, spring hinged temples and pliable temple tips work in combination to give the wearer a comfortable fit optimised to their individual needs.
  • Metal frame safety glasses with an adjustable nose bridge
  • Soft-touch temple tips provide additional comfort
  • Flat temples with spring hinges to wear under a helmet
  • Lens coatings provide both scratch resistance and anti-fogging
  • UV protection with good colour recognition
The sleek wrap-around design provides superior eye coverage and an excellent field of view. Flat temples and a slim nose bridge make this product compatible with a range of protective equipment. They are available in bronze and blue mirror lenses.
Applications include construction and forestry.
Zertifizierungen: EN 166:2001
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge