Cooling plate, 4100
Lieferant: PFM Medical
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
FEAT501003EA 6060 EUR InStock
FEAT501003
Cooling plate, 4100
Kühlbehälter Tischkühler
This cooling plate allows fast and easy cooling of paraffin blocks. The housing is made from stainless steel and the cooling surface is made of anodised aluminium.
  • Easy to clean
  • The temperature is set and controlled electronically
  • User-friendly display and easy to operate
Drain channel in the working surface prevents condensate overflowing after the instrument has been switched off.
Cooling surface (W×D) : 360×246 mm
Working height (H): 67 mm
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge