Organic reference standard, (R)-2-Methyl-3-ureidopropionic acid, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
06997-10MG 06997-50MG
06997-10MGEA 127 EUR InStock
06997-10MG 06997-50MG
Organic reference standard, (R)-2-Methyl-3-ureidopropionic acid, Supelco®
Standards Organische Standards
This metabolite is increased in the urine of patients with beta-ureidopropionase deficiency and can be used to predict patient′s individual phenotypes of enzyme deficiencies in pyrimidine metabolism when associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge