Organic reference standard, N-Succinylglycine, analytical standard, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
01883-10MG 01883-250MG 01883-50MG
01883-10MGEA 94.5 EUR InStock
01883-10MG 01883-250MG 01883-50MG
Organic reference standard, N-Succinylglycine, analytical standard, Supelco®
Standards Organische Standards
Succinylglycine is an acylglycine with a succinyl group as the acyl moiety.
Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders. They are normally produced through the action of glycine N-acyltransferase enzyme.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eade6de597863034c97386080d76bb4b2576bf7&vid=6eade6de597863034c97386080d76bb4b2576bf7&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge