Sigma Pseudo™ corpse scent, drowned victim scent, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
P7184-15CAP P7184-50CAP
P7184-15CAPEA 56.2 EUR InStock
P7184-15CAP P7184-50CAP
Sigma Pseudo™ corpse scent, drowned victim scent, Supelco®
Standards Organische Standards
A valuable training aid for water search. It provides a reliable scent source for 30 to 45 minutes, in still or running water, at depths of 1 to 12 feet. Canine training aids for the detection of corpses.
Verpackung: 15, 50 caps in poly bottle.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge