VWR® SuperCompact, PrEPArative Columns
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
554-4230EA 0 EUR InStock
554-4230 554-4231 554-4232 554-4233 554-4234 554-4235 554-4236 554-4237 554-4238 554-4239
VWR® SuperCompact, PrEPArative Columns
Chromatographiesäulen
These Super Compact preparative columns are made of glass and specially designed for use in aqueous systems.
These columns are containing the following designs such as KPG pipe made of Duran® with transparent sheathing as splinter protection bottom flange and removable compression piston with hand wheel and spindle.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge