Horse Butyrylcholinesterase (from Serum)
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
C7512-1.2KU C7512-12KU C7512-6KU
SIALC7512-1.2KUEA 277 EUR InStock
SIALC7512-1.2KU SIALC7512-12KU SIALC7512-6KU
Horse Butyrylcholinesterase (from Serum)
Enzyme
Butyrylcholinesterase from equine serum has been used in a microcalorimetric study of the inhibition of butyrylcholinesterase by paraoxon. This enzyme has also been used in a study to investigate the synthesis and inhibition of cholinergic enzymes.
  • Lyophilised powder containing buffer salts
Selective inhibition of BChE activity can be used in the detection of organophosphates. Its use in the treatment of organophosphate toxicity has shown clinical potential, as there is a correlation between the blood level of BChE in humans and the degree of protection against toxic nerve agents. There has also been an interest in the roles of cholinesterases with regard to Alzheimer’s disease. Selective inhibition of BChE activity can be used in the detection of organophosphates. Its use in the treatment of organophosphate toxicity has shown clinical potential, as there is a correlation between the blood level of BChE in humans and the degree of protection against toxic nerve agents. There has also been an interest in the roles of cholinesterases with regard to Alzheimer’s disease.
One unit will hydrolyse 1,0 μM of butyrylcholine to choline and butyrate per min at pH 8,0 at 37 °C. The activity obtained using butyrylcholine as substrate is ~2,5 times that obtained using acetylcholine.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge