Sporty S2 safety shoe with wide fit.
- Meets the ESD specification with volume resistance less than 35 megaohms
- Ergonomically designed outsole offers excellent slip resistance
- Water repellent leather
- Smooth-running, dirt- and heat-resistant lacing system
- Reflective details for enhanced visibility
Safety Shoes feature a penetration resistant steel midsole (S3 models) and steel toe cap. Stabilising uvex anti-twist heel cap is designed to prevent ankle twisting. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/blue
Zertifizierungen: EN 61340 (ESD); EN ISO 20345:2011, S2, SRC