Dog Recombinant RC IL-4 (from E. coli)
Lieferant: SHENANDOAH BIOTECHNOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
400-09AF-5UG 400-09AF-25UG 400-09AF-100UG 400-09AF-500UG 400-09AF-1MG
SHBT400-09AF-5UGEA 78.1 EUR InStock
SHBT400-09AF-5UG SHBT400-09AF-25UG SHBT400-09AF-100UG SHBT400-09AF-500UG SHBT400-09AF-1MG
Dog Recombinant RC IL-4 (from E. coli)
Proteine und Peptide
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naive helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
  • High quality
  • Low endotoxin
  • Affordable
Achtung! Bitte beachten: This product is for research use only and is not for use in humans.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge