Live-or-Dye NucFix™ red staining kit
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
32010 32010-T
32010.EA 370 EUR InStock
32010. 32010-T
Live-or-Dye NucFix™ red staining kit
Tests Zelltests
Live-or-Dye NucFix™ Red is a red fluorescent dye to covalently label dead cells, allowing cells with permeable plasma membranes to be detected by flow cytometry or fluorescence microscopy.
  • Nuclear-specific dead cell stain
  • Highly dead-cell specific
  • Covalent labeling with reactive dye
  • Withstand fixation and permeabilization
  • Selective dead-cell staining of mammalian cells, yeast, or gram-negative bacteria
Live-or-Dye NucFix™ Red is a reactive, cell membrane impermeant, red fluorescent dye that specifically stains the nuclei of dead cells. The dye is able to enter into dead cells that have compromised membrane integrity and covalently label the cell nucleus, allowing for clear differentiation of live and dead cells by either microscopy or flow cytometry. Unlike other commonly used nuclear stains such as Propidium Iodide (PI) or DRAQ7™, NucFix™ labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells. The Live-or-Dye NucFix™ Red staining protocol has been optimized to maximize live/dead discrimination with minimal live cell staining, in order to prevent interference with immunostaining. The dye has excitation/emission at 520/593 nm (with DNA).
Bestellinformation: Number of assays is based on 1 mL reaction size (106cells); actual number of assays may vary depending on staining volume.
Lieferumfang: Kit is supplied with components: Live-or-Dye NucFix™ Red Dye and anhydrous DMSO
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge