Signs, COVID-19 reception2, PP, Brady®
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
BRDY306934EA 24.94 EUR InStock
BRDY306934 BRDY306943
Signs, COVID-19 reception2, PP, Brady®
Warn- und Verbotszeichen
Signs are made of surface printed rigid polypropylene material with a slightly textured surface. They are overlaminated with a gloss clear polyester.
  • Ideal for indoor and outdoor applications
  • Well suited for harsh environments
  • Good solvent resistance
  • Rectangular shape
  • Colours: Black and white on red, green, blue, yellow and white
Thickness: 1,501 mm.
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge