Cell culture media, RPMI 1640
Lieferant: PAN BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
P04-17500 P04-18000 P04-16516 P04-18047 P04-17550
PANBP04-17500EA 15.3 EUR InStock
PANBP04-17500 PANBP04-18000 PANBP04-16516 PANBP04-18047 PANBP04-17550
Cell culture media, RPMI 1640
Zellkulturmedien
RPMI 1640 medium was developed for culture of normal and neoplastic leukocytes, but also marrow cells and hybridoma cells.
  • Serum-free media for hybridoma cells
Supplementing RPMI 1640 with varying amounts of FBS, a very good medium for many different cell lines can be obtained.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge