Chemical resistant overalls, AlphaTec® 2000 Ts PLUS, Model 103
Lieferant: ANSELL HEALTH CARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
WH20T-00103-06 WH20T-00103-02 WH20T-00103-03 WH20T-00103-04 WH20T-00103-05 WH20T-00103-07
ANSEWH20T-00103-06EA 498 EUR InStock
ANSEWH20T-00103-06 ANSEWH20T-00103-02 ANSEWH20T-00103-03 ANSEWH20T-00103-04 ANSEWH20T-00103-05 ANSEWH20T-00103-07
Chemical resistant overalls, AlphaTec® 2000 Ts PLUS, Model 103
Einweg-Kleidung
Superior breathable microporous laminate technology, offering Type 4/5/6 protection.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone-free, critical in spray painting applications
  • Tunnelled elasticated wrists and ankles help to minimize the risk of linting and cross contamination
Coveralls feature a collar, single zipper with re-sealable storm flap, and thumb loops. Elasticated waist, tunnelled elasticated wrists and ankles. Garments are manufactured from non-woven microporous polyethylene laminate with serged and taped seams.
Recommended for body painting including surface inspection, cleaning of plant and machines, fibreglass and resin applications, product inspection, and loading and hauling.
Zertifizierungen: Type 4-B; Type 5-B; Type 6-B; EN 1073-2; EN 14126; DIN 32781; EN 1149-5; Category III
Verpackung: Packaged 40 pieces/carton.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge