Labels for BBP33 printers, type B-430
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
133823 133829 133896 153375 153374 153365 153377
BRDY133823EA 158.06 EUR InStock
BRDY133823 BRDY133829 BRDY133896 BRDY153375 BRDY153374 BRDY153365 BRDY153377
Labels for BBP33 printers, type B-430
Etiketten
Glossy metallised permanent polyester (B-435) is designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.
  • Durable permanent acrylic adhesive withstands numerous solvents and variable temperatures
  • Thermal transfer printable
  • Service temperature of –40 to +90 °C
Zertifizierungen: UL approved, CSA accepted.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge