Labels for BBP85 printers, type B-7569
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
120974 120989
BRDY120974EA 163.85 EUR InStock
BRDY120974 BRDY120989
Labels for BBP85 printers, type B-7569
Etiketten
B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
  • Gloss finish
  • Thermal transfer print technology
Ideal for facility identification, floor marking, hazard communication identification, safety identification, warehouse marking.
Zertifizierungen: RoHS compliant with Directive 2011/65/EU and 2015/863, Part does not contain an SVHC, S5609:1986 Section 2 and section 3 approval/compliance.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge