Labels for thermal transfer printers,tType B-727
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
132481 132474 132492
BRDY132481EA 1000.26 EUR InStock
BRDY132481 BRDY132474 BRDY132492
Labels for thermal transfer printers,tType B-727
Etiketten
High temperature polyimide labels to withstand aggressive cleaning processes.
  • Acrylic permanent adhesive
  • Low solvent/chemical resistant
  • Friction resistant
Excellent resistance to flux and wave soldering environments, resistant to all common cleaning methods.
Zertifizierungen: UL recognised; meets the performance requirements of MIL-STD-202G, notice 12, method 215K; free from dibutyltin and dioctyltin.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge