Labels for BBP3x printers, type B-593, rectangular style raised profile
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
118025 118026 118027 118028 118029 118030 118032 118018 118050 118042 118043 118044 118045 118046 118047 118049 118048 118038 118041 118039 118037 118036 118035 118040 118034 118031 118020 118015 118016 118017 118033 118019 118021 118022 118023 118024
BRDY118025EA 322.37 EUR InStock
BRDY118025 BRDY118026 BRDY118027 BRDY118028 BRDY118029 BRDY118030 BRDY118032 BRDY118018 BRDY118050 BRDY118042 BRDY118043 BRDY118044 BRDY118045 BRDY118046 BRDY118047 BRDY118049 BRDY118048 BRDY118038 BRDY118041 BRDY118039 BRDY118037 BRDY118036 BRDY118035 BRDY118040 BRDY118034 BRDY118031 BRDY118020 BRDY118015 BRDY118016 BRDY118017 BRDY118033 BRDY118019 BRDY118021 BRDY118022 BRDY118023 BRDY118024
Labels for BBP3x printers, type B-593, rectangular style raised profile
Etiketten
Raised panel adhesive-taped polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches and internal connection points.
  • Can be used as rating and serial plates using alphanumerics that require nameplate quality
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
  • Service temperature of −20 to 100 °C
Zertifizierungen: UL approved.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge