Labels for BBP33 printers, type B-437
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
133846 133847 133888 133889 133980 133981 133990 133997 133791
BRDY133846EA 417.14 EUR InStock
BRDY133846 BRDY133847 BRDY133888 BRDY133889 BRDY133980 BRDY133981 BRDY133990 BRDY133997 BRDY133791
Labels for BBP33 printers, type B-437
Etiketten
Durable B-437 topcoated polyvinylfluoride material is used most frequently in wire and cable identification applications where self-extinguishing features are required.
  • White or yellow labels feature a matte finish and are thermal transfer printable
  • Service temperature, –70 to 135 °C
This material utilises a permanent acrylic adhesive and has excellent resistance to water, oil and other common fluids.
Zertifizierungen: Meets the requirements of MIL-M-87958, pressure sensitive adhesive wire or cable marker and Identification specification; UL approved; CSA accepted.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge