Multielement Standard Solution 5 for ICP
Lieferant: Honeywell Research Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Fluka™
HONCE2232-100MLEA 115.67 EUR InStock
HONCE2232-100ML
Multielement Standard Solution 5 for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Ca, Fe, K, Na, 10 mg/L each of Ag, Al, Ba, Be, Bi, Cd, Co, Cr, Cs, Cu, Ga, In, Li, Mg, Mn, Mo, Ni, Pb, Rb, Sr, Tl, V, Zn in HNO₃ 10%
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge