High performance mechanic gloves, A710
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
A710BKRXL A710BKRM A710BKRL A710BKRXXL
PRTWA710BKRXLEA 158 EUR InStock
PRTWA710BKRXL PRTWA710BKRM PRTWA710BKRL PRTWA710BKRXXL
High performance mechanic gloves, A710
Handschuhe
Designed and created with heavy duty protection, the Tradesman offers a high level of comfort when working.
  • Additional knuckle padding for extra protection
  • Superior strength and comfort
  • Reinforcement stitching on palms and fingers
  • Hook and loop strap for safe secure fit
  • ANSI abrasion level 3
Built in sweat wipe, knuckle padding and reinforcement stitching on the palms, fingers and thumbs.
Highly durable synthetic leather with spandex, rubber, elastane and neoprene.
Zertifizierungen: Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, EN388:2016 +A1:2018 - 1.1.3.1.X, EN 420
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge