Safety shoes, lace-up, Compositelite™ WORK FC44
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
FC44BKR44 FC44BKR45 FC44BKR46 FC44BKR47 FC44BKR42 FC44BKR43 FC44BKR40 FC44BKR41 FC44BKR38 FC44BKR39 FC44BKR48
PRTWFC44BKR44EA 68.2 EUR InStock
PRTWFC44BKR44 PRTWFC44BKR45 PRTWFC44BKR46 PRTWFC44BKR47 PRTWFC44BKR42 PRTWFC44BKR43 PRTWFC44BKR40 PRTWFC44BKR41 PRTWFC44BKR38 PRTWFC44BKR39 PRTWFC44BKR48
Safety shoes, lace-up, Compositelite™ WORK FC44
Schuhe und Stiefel
Metal-free, anti-static, lightweight safety shoe made from full grain leather.
  • Energy absorbing seat region
  • Composite toecap (200 J)
  • Pierce resistant composite midsole
  • Water resistant upper
Hardwearing dual density PU/TPU outsole with superb slip and oil resistance.
Zertifizierungen: EN ISO 20345:2011, S3, SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge