Safety shoes, lace-up, Steelite™ ULTRA Safety Loire Low Cut Trainer, FW36
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
FW36GRR42 FW36GRR43 FW36GRR44 FW36GRR45 FW36GRR46 FW36GRR47 FW36GRR39 FW36GRR40 FW36GRR41
PRTWFW36GRR42EA 66.7 EUR InStock
PRTWFW36GRR42 PRTWFW36GRR43 PRTWFW36GRR44 PRTWFW36GRR45 PRTWFW36GRR46 PRTWFW36GRR47 PRTWFW36GRR39 PRTWFW36GRR40 PRTWFW36GRR41
Safety shoes, lace-up, Steelite™ ULTRA Safety Loire Low Cut Trainer, FW36
Schuhe und Stiefel
Anti-static safety footwear made from cow suede leather plus a dual density light weight EVA/rubber sole, resistant to heat (300 °C).
  • Protective steel toecap
  • Steel midsole
  • Energy absorbing seat region
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
Zertifizierungen: AS 2210.3; EN ISO 20345:2011, SRC, S1P, HRO.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge