Limited volume inserts, for ND9 and ND11 screw neck and crimp top vials
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SCOT4225800EA 17.27 EUR InStock
SCOT4225800
Limited volume inserts, for ND9 and ND11 screw neck and crimp top vials
Limited volume inserts are used in combination with standard 12×32 mm chromatography vials to facilitate the analysis of small sample volumes.
  • Flat or conical bottom
Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge