Scoop sampling system, PS, double-bagged, Sterileware®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
H36922-1010 H36924-1010
231-0639EA 176 EUR InStock
231-0639 231-0640
Scoop sampling system, PS, double-bagged, Sterileware®, SP Bel-Art
Schaufeln
The long handle increases the distance between the hand and sample to reduce contamination potential. Double-bagging reduces contamination in multi-zone cleanrooms, while the included lid eliminates the cost of separate sterile bags or containers.
  • Snap-on lid allows scoop to be used as a container for transport; lock-tab can be bent under for a tighter seal
  • Sampling data can be marked on the lid with wax pencil, pre-marked label or labelling tape to facilitate tracking
  • Scoops remain upright when placed on a flat surface; can be used as weighing boats
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10⁻⁶
  • Lot stamped for accurate lot tracing, and shipped with a certificate of processing for sterilisation
Bagged twice for extra protection from contamination. 10 individually packed scoops are sealed in an outer bag to create multiple layers of protection against contaminants and debris.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge