Low thermal mass LTM II columns, J&W
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
G3900-65005 G3900-65006
HEWLG3900-65005EA 1878 EUR InStock
HEWLG3900-65005 HEWLG3900-65006
Low thermal mass LTM II columns, J&W
Chromatographiesäulen
Features and benefits for the low thermal mass column modules, or the LTM II.
  • LTM II technology is available for our popular 7890 and 6890 series GC systems
  • The capacity to run up to four column modules simultaneously - with four different temperature programs - to maximize your productivity
  • Rapid temperature programming rates for higher analysis speeds
  • Faster heating and cooling times - as low as one minute or less - for more rapid analytical cycle times and less downtime
  • Excellent retention time repeatability and performance comparable to conventional GC
JW LTM column modules combine a high quality fused silica capillary column with heating and temperature sensing components for a low thermal mass column assembly. The LTM column module contains a patented design which heats and cools the column very efficiently for significantly shorter analytical cycle times compared to conventional air-bath GC oven techniques, while simultaneously using less power. The LTM II transfer line module provides the interface between the standard LTM II 5 in column module and the GC oven. The transfer line module has two heated tubes (transfer lines) through which the column leads pass from the LTM column module into the oven. These transfer lines are temperature programmable to prevent cold spots in the sample path between the GC oven and the LTM column assembly. Each LTM column module attaches to a transfer line module, and the resulting module assembly inserts into slots in the LTM oven door.
LTM II low thermal mass column modules for 7890 series GC systems are available in a wide variety of wall coated open tubular (WCOT) and select porous layer open tubular (PLOT) column configurations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge