Lloyd and McCown's woody plant medium with vitamins
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PML449B-10L PML449C-50L
APOSPML449B-10LEA 68 EUR InStock
APOSPML449B-10L APOSPML449C-50L
Lloyd and McCown's woody plant medium with vitamins
Zellkulturmedien
Contains the WPM macro- and micronutrients and vitamins as described by Lloyd and McCown (1981). Plant tissue culture tested.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge