GC columns, J&W DB-ProSteel
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
145-1011 145-1334 145-2814 145-7032
HEWL145-1011EA 568 EUR InStock
HEWL145-1011 HEWL145-1334 HEWL145-2814 HEWL145-7032
GC columns, J&W DB-ProSteel
Chromatographiesäulen
DB-ProSteel metal columns are virtually unbreakable, and are available in a wide variety of stationary phases.
  • Bonded and cross-linked
  • Ideal for high temperature analysis and process applications
Agilent JW DB-ProSteel metal columns are deactivated with a special process treatment formula to provide good inertness. The columns can be custom-wound on request for small GC ovens. DB-ProSteel has the same outer diameter as standard megabore columns of 0,53 mm Int.Ø so no special ferrules are required for proper installation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge