GC columns, J&W DB-Dioxin
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
122-2461 122-2462
HEWL122-2461EA 1329 EUR InStock
HEWL122-2461 HEWL122-2462
GC columns, J&W DB-Dioxin
Chromatographiesäulen
DB-Dioxin is specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDD) and dibenzofurans (PCDF).
  • Specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDDs) and dibenzofurans (PCDFs)
  • Resolves 2,3,7,8-TCDD and 2,3,7,8-TCDF from all other isomers in one run
  • Low bleed
  • Bonded and cross-linked
  • Solvent rinsable
A 100-psi regulator is required to reach optimum carrier gas velocity.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge