ICP-OES torches for PerkinElmer
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
8003-0344 8003-0345 8003-0346 8003-0347 8003-0348
HEWL8003-0344EA 0 EUR InStock
HEWL8003-0344 HEWL8003-0345 HEWL8003-0346 HEWL8003-0347 HEWL8003-0348
ICP-OES torches for PerkinElmer
Chromatographie-Zubehör Chromatographie-Ersatzteile
These torches for PerkinElmer ICP-OES are available in a wide range of styles from 0 to 3 slots, depending on the instrument and your application needs.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Torches without slots are recommended for organic solvents. One-slot torches are the standard torch for aqueous solutions. Three-slot torches are typically used for organic analysis.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge