J&W DB-ALC1 and DB-ALC2 GC columns
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
123-9134 123-9234 125-9134 125-9234 123-9234E 125-9134E
HEWL123-9134EA 809 EUR InStock
HEWL123-9134 HEWL123-9234 HEWL125-9134 HEWL125-9234 HEWL123-9234E HEWL125-9134E
J&W DB-ALC1 and DB-ALC2 GC columns
Chromatographiesäulen
DB-ALC1 and DB-ALC2 results are complete in just two minutes, reducing run time by one-third compared to other blood alcohol analysis columns.
  • Reliable blood alcohol analysis
  • Optimised primary and confirmation column pair for U.S. blood alcohol analysis
  • Faster GC run times
  • Available in 0,32 and 0,53 mm Int.Ø
Today’s automated static headspace systems use multi-sample equilibration ovens. Its unique polarity, DB-ALC2 provides better resolution of the key ethanol/acetone peaks than other available blood alcohol confirmation capillary columns. These columns are suitable for both headspace analysis and direct injections.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge