Columns, PlusPore
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PL1913-5300 PL1913-5325 PL1913-5500 PL1913-5520
HEWLPL1913-5300EA 2575 EUR InStock
HEWLPL1913-5300 HEWLPL1913-5325 HEWLPL1913-5500 HEWLPL1913-5520
Columns, PlusPore
Chromatographiesäulen
The PlusPore family offers a major resolution improvement over mixed medias and wider range than single pore columns.
  • Separation is maximised over four common MW ranges
  • Small particles and large pore volumes give very fast, high resolution
  • High speed chromatography offers 6 min separations for common samples
The presence of multiple pore sizes on each particle maximises the separation efficiency while still offering a wide MW range with no dislocations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge