GC columns, J&W DB-225ms
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
122-2912 122-2932 122-2962 123-2932
HEWL122-2912EA 517 EUR InStock
HEWL122-2912 HEWL122-2932 HEWL122-2962 HEWL123-2932
GC columns, J&W DB-225ms
Chromatographiesäulen
DB-225ms is excellent for FAME separations and is bonded, cross-linked, and operates from 40 to 240 °C.
  • Virtually equivalent to (50%-Cyanopropylphenyl)-methylpolysiloxane
  • Mid/high polarity
  • Excellent for separations of cis- and trans-fatty acid methyl esters (FAMEs)
  • Low bleed
  • Bonded and cross-linked
J&W DB-225ms columns deliver excellent low bleed performance in a variety of applications and are QC tested to exacting industry standards in terms of accuracy and reproducibility of results. These columns are bonded, cross-linked and low-bleed, and have been precision-engineered to be perfectly suited for efficient separations of cis- and trans-fatty acid methyl esters. The mid- to high-polarity DB-225ms columns are operational from 40 to 240 °C and are also solvent rinsable.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge