Mouse recombinant IL-15 E. coli
Lieferant: BIOLEGEND INC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
566301 566304
BLEG566301EA 134 EUR InStock
BLEG566301 BLEG566304
Mouse recombinant IL-15 E. coli
Proteine und Peptide
For maximum results, quick spin vial prior to opening. Reconstitute in sterile water to a concentration of 0,1 mg/ml including a carrier protein such as 1% BSA or HSA or 10% FBS.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge