ESD safety ankle boots, lace-up, Henny XXTL Mid
Lieferant: ELTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
746581/35 746581/36 746581/37 746581/38 746581/39 746581/40 746581/41 746581/42
ELTN746581/35EA 98.9 EUR InStock
ELTN746581/35 ELTN746581/36 ELTN746581/37 ELTN746581/38 ELTN746581/39 ELTN746581/40 ELTN746581/41 ELTN746581/42
ESD safety ankle boots, lace-up, Henny XXTL Mid
Schuhe und Stiefel
Ladies' safety boot is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • PU/PU sole WELLMAXX TRAINERS LADY
  • Steel toe cap
Nubuck leather/hydrophobised CORDURA® textile material construction with fabric lining for natural ventilation. Full-length inlay ESD sole, PRO Lady Y. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Zertifizierungen: EN ISO 20345 S3 SRC, Form B.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge