ESD safety shoes, lace-up, Broker XXB Low
Lieferant: ELTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
729310/39 729310/40 729310/41 729310/42 729310/43 729310/44 729310/45 729310/46 729310/47 729310/48 729310/49
ELTN729310/39EA 97.9 EUR InStock
ELTN729310/39 ELTN729310/40 ELTN729310/41 ELTN729310/42 ELTN729310/43 ELTN729310/44 ELTN729310/45 ELTN729310/46 ELTN729310/47 ELTN729310/48 ELTN729310/49
ESD safety shoes, lace-up, Broker XXB Low
Schuhe und Stiefel
ESD approved safety shoes and occupational shoes do not only protect the personnel, but also provide protection for components and assemblies at risk from electrostatic energy.Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • MONO-PU sole WELLMAXX BUSINESS
  • Steel toe cap
  • Leather-free
Microfibre construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Soft fleece ESD insole. Sole core made of Infinergy® by BASF.
Zertifizierungen: EN ISO 20345 S1 SRC, form A.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge