Safety spectacle, Galactus™
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
ASA480-0A7-600 ASA480-0B7-600
JSPAASA480-0A7-600EA 23.8 EUR InStock
JSPAASA480-0A7-600 JSPAASA480-0B7-600
Safety spectacle, Galactus™
Schutzbrillen
Adaptable and ultra-light safety spectacle with base ten lens.
  • Stylish premium safety eyewear
  • Ultra-light TPR frame
  • Adaptable nose bridge
  • Base ten lens
  • Soft touch arms
Galactus™ premium safety eyewear features a base 10 curved lens which follows facial contours perfectly, a unique mouldable nose bridge system that can be adjusted to accommodate all high/low/thin or wide nose bridge profiles, and a frame that utilises a fusion of polymer and TPE to be both lightweight and super strong.
Soft touch arms with elastomer-injected sleeve for more comfort and better adhesion. Polymer inner structure for added flexibility and comfort. The base ten lens provides maximum field of view and more protective coverage.
Zertifizierungen: EN 166.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge