Erlenmeyer flasks with plain bottom, sterile, single-use, Thermo Scientific™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
4117-0125 4117-0500
NUNC4117-0125EA 216 EUR InStock
NUNC4117-0125 NUNC4117-0500
Erlenmeyer flasks with plain bottom, sterile, single-use, Thermo Scientific™
Kolben Erlenmeyerkolben
The Thermo Scientific™ single-use PETG Erlenmeyer flasks with plain bottom are ideal for suspension of cell culture, media preparation, mixing and storage.
  • Manufactured with high quality virgin resins
  • Sterilised by gamma irradiation per ISO 11137-2
  • Non-pyrogenic, non-cytotoxic
  • DNase/Rnase, BSE/TSE and human DNA free
  • Molded-in graduation for accurate volume assessment
Made of crystal clear PETG, these flasks are robust and break resistant. Vent cap for continuous gas exchange while maintaining sterility and preventing leakage.
Zertifizierungen: Compliant with California Prop 65.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge