Thermo Scientific Nalgene Conical Centrifuge Bottles, 175 mL
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MFLX06106-08EA 0 EUR InStock
MFLX06106-08 MFLX06107-28
Thermo Scientific Nalgene Conical Centrifuge Bottles, 175 mL
Flaschen Zentrifugenflaschen
Get better separation of large samples.
  • Measure more precisely with molded-in graduations
  • Leakproof
The bottles are available in PPCO or PC. Feature molded-in graduations and PP sealing cap. Must use adapter 06106-78 to fit most rotors.
Lieferumfang: Includes leakproof PP cap.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge