High-Efficiency Pleated Polypropylene Filter Cartridges
Lieferant: Pentair
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MFLX29830-01EA 282.25 EUR InStock
MFLX29830-01 MFLX29830-00
High-Efficiency Pleated Polypropylene Filter Cartridges
Filter Filterkartuschen
Pleated filter gives you more surface area for higher flowrate and longer life.
  • Constructed of pleasted polypropylene
High-Efficiency Pleated Polypropylene (PP) Filter Cartridges provide more surface area for higher flow rate and longer life. Designed for food, pharmaceutical, semiconductor, and other demanding process industries. All PP construction and gradient density microfiber media provide excellent micron ratings and high contaminant holding capacities. All components are biologically safe and are FDA compliant. Maximum temperature is 122 °F (50 °C). Cartridges have Buna N gasket/O-ring. Note: For optimal performance, change cartridge at 25 psid.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge