Serum-free/protein-free medium, 1X, Corning® insectagro®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Corning®
MDTC13-410-CVEA 211.37 EUR InStock
MDTC13-410-CV
Serum-free/protein-free medium, 1X, Corning® insectagro®
Zellkulturmedien
Corning® insectagro® Sf9 is formulated to support the propagation of Sf9 insect cells in culture, and can also be used for SF21 cells.
Sf9 cells are cultured in non-humidified, non-CO₂ incubators at 27 °C (room temperature) and display both monolayer and suspension culture qualities. With their fast doubling times of 10 to 22 hours, Sf9 cells are easily scaled up to large cultures using bioreactors.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge