Burkert® Solenoid-Operated VDC Valves
Lieferant: Burkert
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MFLX98621-40EA 316.24 EUR InStock
MFLX98621-40 MFLX98621-42 MFLX98619-00
Burkert® Solenoid-Operated VDC Valves
Ventile Solenoidventile
Fast-acting long life solenoid valves.
  • Constructed of durable stainless steel (SS) and fluoropolymer elastomer (FPM)
Choose from two- or three-way valves, with the port, orifice, and voltage your applciation requires.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge