Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
76804-13 76804-31 76804-40 76804-41 76804-43
MFLX76804-13EA 2830 EUR InStock
MFLX76804-13 MFLX76804-31 MFLX76804-40 MFLX76804-41 MFLX76804-43
Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Pumpen Exzenterschneckenpumpen
Excellent chemical resistance, pulseless flow, and low shear.
  • Gentle low-shear fluid transfer capable of pumping small particles without damaging product
  • Pumps can handle a variety of polymers and chemicals—ideal for pharmaceutical and water treatment applications
  • Sanitary models approved for 3A sanitary applications and can be cleaned in place
A
Pump heads can be mounted on a NEMA 56C frame motor. Pump heads can also be directly connected or belt driven by an AC or DC motor, gasoline engine (min. 2 hp), or air motor.
B
Pumps are close-coupled to an AC motor with an integral VFD controller. The VFD controller features a “hands-off-auto” switch, providing speeds from 30 to 600 rpm.
C
Pumps feature a ½-hp motor with 230/460 VAC input power. This configuration has a mechanical variable-speed drive that adjusts from 0 to 1000 rpm.
Options are broken up into an industrial and a sanitary categories. Other elastomers are avialble, please contact an Applications Specialts for additional information.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge